Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Nucleoside-Analog-1 | 876707-99-2 | 95.0% | C9H9N5O5 | 100 MG
SDP

Catalog No. 5000433460
Encompass_Preferred

Nucleoside-Analog-1 is a click chemistry reagent containing an azide group. It can participate in copper-catalyzed azide-alkyne cycloaddition reactions (CuAAc) with molecules that have alkyne groups. Additionally, it can undergo strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups.

  • Click chemistry reagent with an azide group
  • Participates in copper-catalyzed azide-alkyne cycloaddition reactions
  • Undergoes strain-promoted alkyne-azide cycloaddition reactions
  • Purity of 95.0%
  • Molecular formula C9H9N5O5
  • Appearance: White to off-white solid
  • Related to anti-infection and cell cycle/DNA damage pathways
  • Associated with disease areas like cancer, infection, and hepatitis C virus infection

Catalog No. 50-004-33460 Supplier Medchemexpress LLC Supplier No. HY77651100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Nucleoside-Analog-1 is a click chemistry reagent containing an azide group. It can participate in copper-catalyzed azide-alkyne cycloaddition reactions (CuAAc) with molecules that have alkyne groups. Additionally, it can undergo strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups.

  • Click chemistry reagent with an azide group
  • Participates in copper-catalyzed azide-alkyne cycloaddition reactions
  • Undergoes strain-promoted alkyne-azide cycloaddition reactions
  • Purity of 95.0%
  • Molecular formula C9H9N5O5
  • Appearance: White to off-white solid
  • Related to anti-infection and cell cycle/DNA damage pathways
  • Associated with disease areas like cancer, infection, and hepatitis C virus infection

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.