Learn More
Medchemexpress LLC Nucleoside-analog-1 | 876707-99-2 | 95.0% | C9H9N5O5 | 2 MG
SDP
Catalog No. 5000435115
Encompass_Preferred
Nucleoside-Analog-1 is a click chemistry reagent. It contains an Azide group and can undergo copper-catalyzed azide-alkyne cycloaddition reaction (CuAAc) with molecules containing Alkyne groups. It can also undergo strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups.
  • Contains an azide group
  • Can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • Compatible with molecules containing alkyne groups
  • Can undergo strain-promoted alkyne-azide cycloaddition (SPAAC)
  • Compatible with molecules containing DBCO or BCN groups
Catalog No. 50-004-35115 Supplier Medchemexpress LLC Supplier No. HY776512MG
Nucleoside-Analog-1 is a click chemistry reagent. It contains an Azide group and can undergo copper-catalyzed azide-alkyne cycloaddition reaction (CuAAc) with molecules containing Alkyne groups. It can also undergo strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups.
  • Contains an azide group
  • Can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • Compatible with molecules containing alkyne groups
  • Can undergo strain-promoted alkyne-azide cycloaddition (SPAAC)
  • Compatible with molecules containing DBCO or BCN groups