Learn More
Medchemexpress LLC Nucleoside-Analog-1 | 876707-99-2 | 95.0% | C9H9N5O5 | 25 MG
SDP
Catalog No. 5000359001
Encompass_Preferred
Nucleoside-Analog-1 is a click chemistry reagent containing an Azide group. It can participate in copper-catalyzed azide-alkyne cycloaddition reaction (CuAAc) with molecules that have Alkyne groups. Additionally, it can undergo strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups.
  • Click chemistry reagent containing an azide group
  • Participates in copper-catalyzed azide-alkyne cycloaddition reaction (CuAAc)
  • Undergoes strain-promoted alkyne-azide cycloaddition (SPAAC) reactions
  • Solid, white to off-white in color
  • Intended for research use only
Catalog No. 50-003-59001 Supplier Medchemexpress LLC Supplier No. 25MGHY77651
Nucleoside-Analog-1 is a click chemistry reagent containing an Azide group. It can participate in copper-catalyzed azide-alkyne cycloaddition reaction (CuAAc) with molecules that have Alkyne groups. Additionally, it can undergo strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups.
  • Click chemistry reagent containing an azide group
  • Participates in copper-catalyzed azide-alkyne cycloaddition reaction (CuAAc)
  • Undergoes strain-promoted alkyne-azide cycloaddition (SPAAC) reactions
  • Solid, white to off-white in color
  • Intended for research use only