Learn More
Medchemexpress LLC Nucleoside-Analog-2 | 876708-01-9 | ≥95.0% | C9H11N5O6 | 1 ML
SDP
Catalog No. 5000434541
Encompass_Preferred
Nucleoside-Analog-2 is a 4'-Azidocytidine analogue effective against Hepatitis C virus (HCV) replication. It functions as a click chemistry reagent containing an Azide group, enabling it to participate in copper-catalyzed azide-alkyne cycloaddition (CuAAc) with Alkyne groups and strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with DBCO or BCN groups. This product is intended for research use only.
  • 4'-Azidocytidine analogue
  • Active against Hepatitis C virus (HCV) replication
  • Click chemistry reagent
  • Contains an Azide group
  • Can undergo copper-catalyzed azide-alkyne cycloaddition reaction (CuAAc)
  • Can undergo strain-promoted alkyne-azide cycloaddition (SPAAC) reactions
Catalog No. 50-004-34541 Supplier Medchemexpress LLC Supplier No. HY7765210MM/1ML
Nucleoside-Analog-2 is a 4'-Azidocytidine analogue effective against Hepatitis C virus (HCV) replication. It functions as a click chemistry reagent containing an Azide group, enabling it to participate in copper-catalyzed azide-alkyne cycloaddition (CuAAc) with Alkyne groups and strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with DBCO or BCN groups. This product is intended for research use only.
  • 4'-Azidocytidine analogue
  • Active against Hepatitis C virus (HCV) replication
  • Click chemistry reagent
  • Contains an Azide group
  • Can undergo copper-catalyzed azide-alkyne cycloaddition reaction (CuAAc)
  • Can undergo strain-promoted alkyne-azide cycloaddition (SPAAC) reactions