Learn More
Medchemexpress LLC Nucleoside-Analog-2 | 876708-01-9 | ≥95.0% | C9H11N5O6 | 100 MG
SDP
Catalog No. 5000434612
Encompass_Preferred
Nucleoside-Analog-2 is a 4'-Azidocytidine analogue primarily designed to act against Hepatitis C virus (HCV) replication. It functions as a versatile click chemistry reagent, featuring an Azide group, which enables participation in both copper-catalyzed azide-alkyne cycloaddition (CuAAc) and strain-promoted alkyne-azide cycloaddition (SPAAC) reactions.
  • Acts against hepatitis C virus (HCV) replication
  • Functions as a click chemistry reagent
  • Contains an azide group for CuAAc reactions with alkyne-containing molecules
  • Undergoes SPAAC reactions with DBCO or BCN group-containing molecules
  • Intended for research use only
Catalog No. 50-004-34612 Supplier Medchemexpress LLC Supplier No. HY77652100MG
Nucleoside-Analog-2 is a 4'-Azidocytidine analogue primarily designed to act against Hepatitis C virus (HCV) replication. It functions as a versatile click chemistry reagent, featuring an Azide group, which enables participation in both copper-catalyzed azide-alkyne cycloaddition (CuAAc) and strain-promoted alkyne-azide cycloaddition (SPAAC) reactions.
  • Acts against hepatitis C virus (HCV) replication
  • Functions as a click chemistry reagent
  • Contains an azide group for CuAAc reactions with alkyne-containing molecules
  • Undergoes SPAAC reactions with DBCO or BCN group-containing molecules
  • Intended for research use only