Learn More
Medchemexpress LLC Nucleoside-analog-2 | 876708-01-9 | ≥95.0% | C9H11N5O6 | 2MG
SDP
Catalog No. 5000434980
Encompass_Preferred
Nucleoside-Analog-2 is a 4'-Azidocytidine analogue that targets and inhibits the replication of the Hepatitis C virus (HCV). It functions as a click chemistry reagent containing an Azide group, enabling copper-catalyzed azide-alkyne cycloaddition reactions (CuAAc) with Alkyne groups, and strain-promoted alkyne-azide cycloaddition (SPAAC) with DBCO or BCN groups. This product is for research use only.
  • 4'-Azidocytidine analogue
  • Inhibits hepatitis C virus (HCV) replication
  • Click chemistry reagent with an azide group
  • Facilitates CuAAc reactions with alkyne groups
  • Undergoes SPAAC reactions with DBCO or BCN groups
  • For research use only
Catalog No. 50-004-34980 Supplier Medchemexpress LLC Supplier No. HY776522MG
Nucleoside-Analog-2 is a 4'-Azidocytidine analogue that targets and inhibits the replication of the Hepatitis C virus (HCV). It functions as a click chemistry reagent containing an Azide group, enabling copper-catalyzed azide-alkyne cycloaddition reactions (CuAAc) with Alkyne groups, and strain-promoted alkyne-azide cycloaddition (SPAAC) with DBCO or BCN groups. This product is for research use only.
  • 4'-Azidocytidine analogue
  • Inhibits hepatitis C virus (HCV) replication
  • Click chemistry reagent with an azide group
  • Facilitates CuAAc reactions with alkyne groups
  • Undergoes SPAAC reactions with DBCO or BCN groups
  • For research use only