Learn More
Medchemexpress LLC Nucleoside-Analog-2 | 876708-01-9 | ≥95.0% | C9H11N5O6 | 50 MG
SDP
Catalog No. 5000434826
Encompass_Preferred
Nucleoside-Analog-2 is a 4'-Azidocytidine analogue that targets Hepatitis C virus (HCV) replication. It functions as a click chemistry reagent due to its Azide group.
  • Undergoes copper-catalyzed azide-alkyne cycloaddition reactions (CuAAc) with molecules containing Alkyne groups.
  • Can participate in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules possessing DBCO or BCN groups.
  • Targets Hepatitis C virus (HCV) replication.
Catalog No. 50-004-34826 Supplier Medchemexpress LLC Supplier No. HY7765250MG
Nucleoside-Analog-2 is a 4'-Azidocytidine analogue that targets Hepatitis C virus (HCV) replication. It functions as a click chemistry reagent due to its Azide group.
  • Undergoes copper-catalyzed azide-alkyne cycloaddition reactions (CuAAc) with molecules containing Alkyne groups.
  • Can participate in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules possessing DBCO or BCN groups.
  • Targets Hepatitis C virus (HCV) replication.