Learn More
STEMCELL Technologies Uridine, Size: 10 g
SDP
Catalog No. 502069349
Encompass
Uridine is one of the five nucleosides used for RNA synthesis and as a building block for other biomolecules (Tsukimoto). Uridine 5’-diphosphate and uridine 5’-triphosphate are signaling molecules that activate the purinergic receptors (Tsukimoto). Uridine is also used as a medium supplement for culturing primary hippocampal neuron cells (François-Moutal et al.).CANCER RESEARCH· Uridine has been used to reduce 5-fluorouracil toxicity, without affecting its antitumor activity (Pizzorno et al.).
Catalog No. 50-206-9349 Supplier STEMCELL Technologies Supplier No. 1000541
Uridine is one of the five nucleosides used for RNA synthesis and as a building block for other biomolecules (Tsukimoto). Uridine 5’-diphosphate and uridine 5’-triphosphate are signaling molecules that activate the purinergic receptors (Tsukimoto). Uridine is also used as a medium supplement for culturing primary hippocampal neuron cells (François-Moutal et al.).CANCER RESEARCH· Uridine has been used to reduce 5-fluorouracil toxicity, without affecting its antitumor activity (Pizzorno et al.).