Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nucleosides Test Mix in 1 Sodium Formate (varied conc.), Analytical Standard, MilliporeSigma™ Supelco™

Catalog No. 111005041 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-100-5041 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-100-5041 Supplier MilliporeSigma Supelco Supplier No. 47310U
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Nucleosides Test Mix enables the troubleshooting of chromatographic problems, optimization of system efficiency, and evaluation of columns. These test mixes are shipped in amber ampules to prevent photodegradation. The instructions for proper use and interpretation of results are included. The Nucleosides Test Mix can be suitably chosen according to column-specific or application-specific requirements. The identification can be carried out using UV detection. For additional information, HPLC Troubleshooting Guide (Bulletin 826) may be requested.
TRUSTED_SUSTAINABILITY

Specifications

Packaging Ampule
Quantity 1 mL
Grade Analytical Standard
Concentration or Composition (by Analyte or Components) Cytidine 50 μg/mL; Guanosine 25 μg/mL;Inosine 25 μg/mL; 1-Methyladenosine 25 μg/mL; 5-Methylcytidine 100 μg/mL; 2 '-O-Methylcytidine 20 μg/mL; 3-Methylcytidine methosulfate 100 μg/mL; 7-Methylguanosine 25 μg/mL; 5-Methyluridine 50 μg/mL; beta-Pseudouridine 25 μg/mL; 2-Thiocytidine dihydrate 10 μg/mL; Uridine 25 μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.