Learn More
Sigma Aldrich Fine Chemicals Biosciences Nucleosil(R) C18 HPLC Column 5 mum particle size, L x I.D. 25 cm x 4 mm |
Catalog No. NC1766187
Encompass
Nucleosil(R) C18 HPLC Column 5 mum particle size, L x I.D. 25 cm x 4 mm |
Catalog No. NC1766187 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 50166U
Nucleosil(R) C18 HPLC Column 5 mum particle size, L x I.D. 25 cm x 4 mm |