Learn More
Macherey-Nagel HPLC column EC 250/4.6 NUCLEOSIL 100-10 CN
Catalog No. NC0956771
Encompass_Preferred
EC HPLC column - EC 250/4.6 NUCLEOSIL 100-10 CN - length: 250 mm, ID: 4.6 mm - pack of 1
Catalog No. NC0956771 Supplier Macherey-Nagel Supplier No. 720024.46
EC HPLC column - EC 250/4.6 NUCLEOSIL 100-10 CN - length: 250 mm, ID: 4.6 mm - pack of 1