Learn More
Macherey-Nagel EC HPLC column (analytical), NUCLEOSIL 100-5 SB, 5 µm, 125x4.6 mm
Catalog No. NC1805345
Encompass_Preferred
Strongly basic anion exchanger (SAX) with capacity 1 meq/g. Separation mechanism based on strong ion exchange interactions. Based on the original NUCLEOSIL silica gel. It is one of the first spherical silicas for HPLC.
Catalog No. NC1805345 Supplier Macherey-Nagel Supplier No. 720989.46
Strongly basic anion exchanger (SAX) with capacity 1 meq/g. Separation mechanism based on strong ion exchange interactions. Based on the original NUCLEOSIL silica gel. It is one of the first spherical silicas for HPLC.