Learn More
Macherey-Nagel NUCLEOSIL (R) 1000-7 C18 HPLC
Catalog No. NC1837198
Encompass_Preferred
Nucleosil (R) 1000-7 C18 HPLC Column 1000 Angstrom pore size 7 um particle size 4.0 mm diameter (not 4.6 - IMPORTANT) 250 mm length
Catalog No. NC1837198 Supplier Macherey-Nagel Supplier No. 720077.40
Nucleosil (R) 1000-7 C18 HPLC Column 1000 Angstrom pore size 7 um particle size 4.0 mm diameter (not 4.6 - IMPORTANT) 250 mm length