Learn More
Takara Bio NUCLEOSNAP PLASMID MIDI 50PREP
SDP
Catalog No. NC1940445
Encompass
ndotoxins are copurified during plasmid preparations from bacterial lysates. Since they interfere with eukaryotic cell survival, endotoxin reduction is essential prior to cell transfection. Macherey-Nagel has developed a new column format and a novel buffer chemistry to enable vacuum-processed isolation of transfection-grade plasmid DNA in a midiprep format. TAKARA BIO USA INC mfg# 740494.50
Catalog No. NC1940445 Supplier Takara Bio Supplier No. 740494.50
ndotoxins are copurified during plasmid preparations from bacterial lysates. Since they interfere with eukaryotic cell survival, endotoxin reduction is essential prior to cell transfection. Macherey-Nagel has developed a new column format and a novel buffer chemistry to enable vacuum-processed isolation of transfection-grade plasmid DNA in a midiprep format. TAKARA BIO USA INC mfg# 740494.50