Learn More
Active Motif Nucleosome Preparation Kit
SDP
Catalog No. NC1750512
Encompass
Active Motif's Nucleosome Preparation Kit is designed to prepare mono- and oligonucleosomes starting from cells. Nucleosomes comprise the smallest subunit of chromatin and consist of 147 bp of DNA wrapped around an octamer of core histone proteins (H2A, H2B, H3 and H4). Each nucleosome is separated by a linker of DNA. The Nucleosome Preparation Kit uses an enzymatic cocktail to digest the linker DNA. By adjusting the digestion time, it is possible to control the amount of mononucleosomes or oligonucleosomes extracted. Because isolated nucleosomes are more physiologically relevant than histones alone or synthetic peptides, they are ideally suited for use as substrates in the analysis of histone post-translational modifications, enzyme kinetics or inhibitor screening studies.
Catalog No. NC1750512 Supplier Active Motif Supplier No. 53504
Active Motif's Nucleosome Preparation Kit is designed to prepare mono- and oligonucleosomes starting from cells. Nucleosomes comprise the smallest subunit of chromatin and consist of 147 bp of DNA wrapped around an octamer of core histone proteins (H2A, H2B, H3 and H4). Each nucleosome is separated by a linker of DNA. The Nucleosome Preparation Kit uses an enzymatic cocktail to digest the linker DNA. By adjusting the digestion time, it is possible to control the amount of mononucleosomes or oligonucleosomes extracted. Because isolated nucleosomes are more physiologically relevant than histones alone or synthetic peptides, they are ideally suited for use as substrates in the analysis of histone post-translational modifications, enzyme kinetics or inhibitor screening studies.