Learn More
Macherey-Nagel NucleoSpin RNA Midi, 20 preps
Supplier:  Macherey-Nagel 740962.20
Encompass_Preferred
NucleoSpin RNA Midi (20) 20 preps for the isolation of RNA from cells and tissue - NucleoSpin RNA Midi Columns, NucleoSpin RNA Filters Midi, Elution Tubes, buffers, RNase-free rDNase
Catalog No. NC1662306