Learn More
Macherey-Nagel NucleoSpin 96 Blood QuickPure, Efficient DNA Purification, Silica-Membrane Technology, 96-Well Format
Catalog No. NC2608653
Encompass_Preferred
The NucleoSpin 96 Blood QuickPure kit offers an efficient solution for rapid DNA purification from various blood samples, utilizing advanced silica-membrane technology. This kit minimizes hands-on time while ensuring high-purity DNA suitable for sensitive applications like PCR. With a capacity of 384 preps, it is designed for streamlined processes and high yields, setting it apart from other products.
Catalog No. NC2608653 Supplier Macherey-Nagel Supplier No. 740667.4
The NucleoSpin 96 Blood QuickPure kit offers an efficient solution for rapid DNA purification from various blood samples, utilizing advanced silica-membrane technology. This kit minimizes hands-on time while ensuring high-purity DNA suitable for sensitive applications like PCR. With a capacity of 384 preps, it is designed for streamlined processes and high yields, setting it apart from other products.