Learn More
Macherey-Nagel NucleoSpin 96 Tissue Core Kit, 4 x 96 preps
Catalog No. NC1162419
Encompass_Preferred
NucleoSpin 96 Tissue Core Kit (4 x 96) 4 x 96 preps for the isolation of genomic DNA from cells and tissue - NucleoSpin Tissue Binding Plates, buffers, Proteinase K
Catalog No. NC1162419 Supplier Macherey-Nagel Supplier No. 740454.4
NucleoSpin 96 Tissue Core Kit (4 x 96) 4 x 96 preps for the isolation of genomic DNA from cells and tissue - NucleoSpin Tissue Binding Plates, buffers, Proteinase K