Learn More
Macherey-Nagel NucleoSpin Blood, 50 preps
Catalog No. NC0389523
Encompass_Preferred
NucleoSpin Blood (50) 50 preps for the isolation of genomic DNA from blood - NucleoSpin Blood Columns, Collection Tubes, buffers, Proteinase K
Catalog No. NC0389523 Supplier Macherey-Nagel Supplier No. 740951.50
NucleoSpin Blood (50) 50 preps for the isolation of genomic DNA from blood - NucleoSpin Blood Columns, Collection Tubes, buffers, Proteinase K