Learn More
Macherey-Nagel NucleoSpin DNA RapidLyse, 50 preps
Catalog No. NC1569676
Encompass_Preferred
NucleoSpin DNA RapidLyse (50) 50 preps for the isolation of genomic DNA - NucleoSpin DNA RapidLyse Columns, Collection Tubes, buffers, Proteinase K
Catalog No. NC1569676 Supplier Macherey-Nagel Supplier No. 740100.50
NucleoSpin DNA RapidLyse (50) 50 preps for the isolation of genomic DNA - NucleoSpin DNA RapidLyse Columns, Collection Tubes, buffers, Proteinase K