Learn More
Takara Bio NucleoSpin® Gel and PCR Clean-Up, 50 preps
SDP
Catalog No. NC9233808
Encompass
NucleoSpin Gel and PCR Clean-up (50) 50 preps for PCR clean-up and gel extraction - NucleoSpin Gel and PCR Clean-up Columns, Collection Tubes, buffers.
Catalog No. NC9233808 Supplier Takara Bio Supplier No. 740609.50
NucleoSpin Gel and PCR Clean-up (50) 50 preps for PCR clean-up and gel extraction - NucleoSpin Gel and PCR Clean-up Columns, Collection Tubes, buffers.