Learn More
Macherey-Nagel NucleoSpin Gel and PCR Clean-up Midi, 20 preps
Catalog No. NC1478140
Encompass_Preferred
NucleoSpin Gel and PCR Clean-up Midi(20) 20 preps for large scale PCR clean-up and gel extraction, Buffers, NucleoSpin Extract II Midi Columns, Collection Tubes, User Manual
Catalog No. NC1478140 Supplier Macherey-Nagel Supplier No. 740986.20
NucleoSpin Gel and PCR Clean-up Midi(20) 20 preps for large scale PCR clean-up and gel extraction, Buffers, NucleoSpin Extract II Midi Columns, Collection Tubes, User Manual