Learn More
Macherey-Nagel NucleoSpin Gel and PCR Clean-up, 10 preps
Catalog No. NC1239647
Encompass_Preferred
NucleoSpin Gel and PCR Clean-up (10) 10 preps for PCR clean-up and gel extraction - NucleoSpin Gel and PCR Clean-up Columns, Collection Tubes, buffers
Catalog No. NC1239647 Supplier Macherey-Nagel Supplier No. 740609.10
NucleoSpin Gel and PCR Clean-up (10) 10 preps for PCR clean-up and gel extraction - NucleoSpin Gel and PCR Clean-up Columns, Collection Tubes, buffers