Learn More
Macherey-Nagel NucleoSpin miRNA, 10 preps
Catalog No. NC0764933
Encompass_Preferred
NucleoSpin miRNA (10) 10 preps for the isolation of small and large RNA - NucleoSpin RNA Columns, NucleoSpin miRNA Columns, NucleoSpin Filters, NucleoSpin Protein Removal Columns, Collection Tubes, buffers, RNase-free rDNase
Catalog No. NC0764933 Supplier Macherey-Nagel Supplier No. 740971.10
NucleoSpin miRNA (10) 10 preps for the isolation of small and large RNA - NucleoSpin RNA Columns, NucleoSpin miRNA Columns, NucleoSpin Filters, NucleoSpin Protein Removal Columns, Collection Tubes, buffers, RNase-free rDNase