Learn More
Macherey-Nagel MACHEREY-NAGEL NucleoSpin Plasmid Binding Plate, High-Purity Plasmid DNA Isolation, Compatible With Automation - 96 Plates
Catalog No. NC2155722
Encompass_Preferred
The MACHEREY-NAGEL NucleoSpin Plasmid Binding Plate is a specialized 96-well plate for efficient high-throughput isolation of high-copy plasmid DNA from E. coli cultures. It features advanced silica membrane technology which ensures effective purification and high-purity yields suitable for downstream applications like sequencing and PCR. Compatible with vacuum manifolds and automation workstations, it streamlines processes while delivering robust results.
This item is not returnable. View return policy
Catalog No. NC2155722 Supplier Macherey-Nagel Supplier No. 740708.24S
Add to Cart
Add to Cart
The MACHEREY-NAGEL NucleoSpin Plasmid Binding Plate is a specialized 96-well plate for efficient high-throughput isolation of high-copy plasmid DNA from E. coli cultures. It features advanced silica membrane technology which ensures effective purification and high-purity yields suitable for downstream applications like sequencing and PCR. Compatible with vacuum manifolds and automation workstations, it streamlines processes while delivering robust results.