Learn More
Macherey-Nagel NucleoSpin cfDNA XS, 50 preps
Supplier:  Macherey-Nagel 740900.50
Encompass_Preferred
NucleoSpin Plasma XS (50) 50 preps for the isolation of circulating DNA from plasma - NucleoSpin Plasma XS Columns, Collection Tubes, buffers, Proteinase K
Catalog No. NC9959409