Learn More
Macherey-Nagel NucleoSpin Plasmid Buffer Set
Catalog No. NC0572838
Encompass_Preferred
NucleoSpin Plasmid Buffer Set for the isolation of low-copy plasmids, only applicable with NucleoSpin Plasmid kits, Buffers A1, A2, A3, RNase A sufficient for 300 preps
Catalog No. NC0572838 Supplier Macherey-Nagel Supplier No. 740953
NucleoSpin Plasmid Buffer Set for the isolation of low-copy plasmids, only applicable with NucleoSpin Plasmid kits, Buffers A1, A2, A3, RNase A sufficient for 300 preps