Learn More
Takara Bio NucleoSpin Plasmid Transfection-grade | 50 preps
SDP
Catalog No. NC1636337
Encompass
NucleoSpin Plasmid Transfection-grade is a fast miniprep kit that uses a new technology developed by Macherey-Nagel to reduce the level of endotoxins co-purified during plasmid preparations from bacterial lysates. Since endotoxins interfere with eukaryotic cell survival, endotoxin reduction is essential prior to cell transfection.
Catalog No. NC1636337 Supplier Takara Bio Supplier No. 740490.50
NucleoSpin Plasmid Transfection-grade is a fast miniprep kit that uses a new technology developed by Macherey-Nagel to reduce the level of endotoxins co-purified during plasmid preparations from bacterial lysates. Since endotoxins interfere with eukaryotic cell survival, endotoxin reduction is essential prior to cell transfection.