Learn More
Takara Bio NucleoSpin RNA Blood Midi | 20 Preps
SDP
Catalog No. NC1260703
Encompass
The NucleoSpin RNA Blood Midi kit enables efficient recovery of high-quality RNA from up to 1.3 ml of whole blood using a simple and convenient procedure that can be performed at room temperature. The kit protocol is compatible with common blood collection tubes and anticoagulants—including EDTA, citrate, and heparin—and enables processing of 6 samples in 75 minutes.
Catalog No. NC1260703 Supplier Takara Bio Supplier No. 740210.20
The NucleoSpin RNA Blood Midi kit enables efficient recovery of high-quality RNA from up to 1.3 ml of whole blood using a simple and convenient procedure that can be performed at room temperature. The kit protocol is compatible with common blood collection tubes and anticoagulants—including EDTA, citrate, and heparin—and enables processing of 6 samples in 75 minutes.