Learn More
Takara Bio NUCLEOSPIN RNA FLTR PLTE PK/4
SDP
Supplier:  Takara Bio 740711
Encompass
NucleoSpin RNA Filter Plates (4) 96-well plates for filtration of cell and tissue homogenates pack of 4
Catalog No. NC2397461