Learn More
Macherey-Nagel NucleoSpin RNA Plant, 250 preps
Catalog No. NC1409717
Encompass_Preferred
NucleoSpin RNA Plant (250) 250 preps for the isolation of RNA from plant - NucleoSpin RNA Plant Columns, NucleoSpin Filters, Collection Tubes, buffers, RNase-free rDNase
Catalog No. NC1409717 Supplier Macherey-Nagel Supplier No. 740949.250
Add to Cart
Add to Cart
NucleoSpin RNA Plant (250) 250 preps for the isolation of RNA from plant - NucleoSpin RNA Plant Columns, NucleoSpin Filters, Collection Tubes, buffers, RNase-free rDNase