Learn More
Macherey-Nagel NucleoSpin RNA Plant, 50 preps
Catalog No. NC0648387
Encompass_Preferred
NucleoSpin RNA Plant (50) 50 preps for the isolation of RNA from plant - NucleoSpin RNA Plant Columns, NucleoSpin Filters, Collection Tubes, buffers, RNase-free rDNase
Catalog No. NC0648387 Supplier Macherey-Nagel Supplier No. 740949.50
NucleoSpin RNA Plant (50) 50 preps for the isolation of RNA from plant - NucleoSpin RNA Plant Columns, NucleoSpin Filters, Collection Tubes, buffers, RNase-free rDNase