Learn More
Macherey-Nagel NucleoSpin RNA Plus Mini Kit, Isolates High-Purity RNA, 50 Preps
Catalog No. NC1749027
Encompass_Preferred
The MACHEREY NAGE NucleoSpin RNA Plus Mini kit provides an ultrafast method for high-purity RNA isolation from diverse biological samples, ensuring genomic DNA removal without rDNase. Its silica membrane technology offers reliable recovery, typical RNA Integrity Numbers (RIN) exceeding 9, and an A260/A280 ratio greater than 1.9. Suitable for RT-PCR and RNA sequencing, it features a 50 prep capacity and a preparation time of 20 min for 6 preps.
Catalog No. NC1749027 Supplier Macherey-Nagel Supplier No. 740984.50
The MACHEREY NAGE NucleoSpin RNA Plus Mini kit provides an ultrafast method for high-purity RNA isolation from diverse biological samples, ensuring genomic DNA removal without rDNase. Its silica membrane technology offers reliable recovery, typical RNA Integrity Numbers (RIN) exceeding 9, and an A260/A280 ratio greater than 1.9. Suitable for RT-PCR and RNA sequencing, it features a 50 prep capacity and a preparation time of 20 min for 6 preps.