Learn More
Macherey-Nagel NucleoSpin RNA Plus XS, Micro kit for RNA purification with DNA removal column, 10 preps per pack
Catalog No. NC1882501
Encompass_Preferred
NucleoSpin RNA Plus XS, Micro kit for RNA purification with DNA removal column. XS spin kit for ultrafast and convenient RNA isolation without rDNase digestion.
Catalog No. NC1882501 Supplier Macherey-Nagel Supplier No. 740990.10
NucleoSpin RNA Plus XS, Micro kit for RNA purification with DNA removal column. XS spin kit for ultrafast and convenient RNA isolation without rDNase digestion.