Learn More
Macherey-Nagel NucleoSpin RNA Plus XS, 50 preps
Catalog No. NC1498148
Encompass_Preferred
NucleoSpin RNA Plus XS (50) 50 preps for the isolation of total RNA - NucleoSpin RNA Plus XS Columns, NucleoSpin gDNA Removal XS Columns, Collection Tubes, buffers
Catalog No. NC1498148 Supplier Macherey-Nagel Supplier No. 740990.50
NucleoSpin RNA Plus XS (50) 50 preps for the isolation of total RNA - NucleoSpin RNA Plus XS Columns, NucleoSpin gDNA Removal XS Columns, Collection Tubes, buffers