Learn More
Macherey-Nagel NucleoSpin RNA/Protein, 50 preps
Catalog No. NC1081772
Encompass_Preferred
NucleoSpin RNA/Protein (50) 50 preps for the isolation of RNA and protein from one lysate - NucleoSpin RNA/Protein Columns, NucleoSpin Filters, Collection Tubes, buffers, RNase-free rDNase
Catalog No. NC1081772 Supplier Macherey-Nagel Supplier No. 740933.50
NucleoSpin RNA/Protein (50) 50 preps for the isolation of RNA and protein from one lysate - NucleoSpin RNA/Protein Columns, NucleoSpin Filters, Collection Tubes, buffers, RNase-free rDNase