Learn More
Macherey-Nagel NucleoSpin RNA XS, 50 preps
Catalog No. NC0389511
Encompass_Preferred
NucleoSpin RNA XS (50) 50 preps for the isolation of total RNA - NucleoSpin RNA XS Columns, NucleoSpin Filters, Collection Tubes, buffers, RNase-free rDNase, Carrier RNA, Reducing Agent TCEP
Catalog No. NC0389511 Supplier Macherey-Nagel Supplier No. 740902.50
NucleoSpin RNA XS (50) 50 preps for the isolation of total RNA - NucleoSpin RNA XS Columns, NucleoSpin Filters, Collection Tubes, buffers, RNase-free rDNase, Carrier RNA, Reducing Agent TCEP