Learn More
Macherey-Nagel NucleoSpin Soil, 250 preps
Catalog No. NC0706445
Encompass_Preferred
NucleoSpin Soil (250) 250 preps for the isolation of DNA from soil and sediment - NucleoSpin Soil Columns, NucleoSpin Bead Tubes, NucleoSpin Inhibitor Removal Columns, Collection Tubes, buffers
Catalog No. NC0706445 Supplier Macherey-Nagel Supplier No. 740780.250
NucleoSpin Soil (250) 250 preps for the isolation of DNA from soil and sediment - NucleoSpin Soil Columns, NucleoSpin Bead Tubes, NucleoSpin Inhibitor Removal Columns, Collection Tubes, buffers