Learn More
Macherey-Nagel NucleoSpin Tissue, 250 preps
Catalog No. NC0389454
Encompass_Preferred
NucleoSpin Tissue (250) 250 preps for the isolation of genomic DNA from tissue - NucleoSpin Tissue Columns, Collection Tubes, buffers, Proteinase K
Catalog No. NC0389454 Supplier Macherey-Nagel Supplier No. 740952.250
NucleoSpin Tissue (250) 250 preps for the isolation of genomic DNA from tissue - NucleoSpin Tissue Columns, Collection Tubes, buffers, Proteinase K