Learn More
Macherey-Nagel NucleoSpin Tissue, 50 preps
Catalog No. NC0389409
Encompass_Preferred
NucleoSpin Tissue (50) 50 preps for the isolation of genomic DNA from tissue - NucleoSpin Tissue Columns, Collection Tubes, buffers, Proteinase K
Catalog No. NC0389409 Supplier Macherey-Nagel Supplier No. 740952.50
NucleoSpin Tissue (50) 50 preps for the isolation of genomic DNA from tissue - NucleoSpin Tissue Columns, Collection Tubes, buffers, Proteinase K