Learn More
Macherey-Nagel NucleoSpin Virus, 50 preps
Catalog No. NC1410232
Encompass_Preferred
NucleoSpin Virus (50) 50 preps for the isolation of viral RNA / DNA from serum and plasma - NucleoSpin Virus XS Columns, Collection Tubes, buffers, Liquid Proteinase K, Carrier RNA
Catalog No. NC1410232 Supplier Macherey-Nagel Supplier No. 740983.50
NucleoSpin Virus (50) 50 preps for the isolation of viral RNA / DNA from serum and plasma - NucleoSpin Virus XS Columns, Collection Tubes, buffers, Liquid Proteinase K, Carrier RNA