Learn More
Macherey-Nagel NucleoVac 96
Catalog No. NC1229304
Encompass_Preferred
NucleoVac 96 Vacuum Manifold vacuum manifold for use of NucleoSpin 8/96 kits under vacuum - manifold base, manifold lid, waste containers, spacer set
Catalog No. NC1229304 Supplier Macherey-Nagel Supplier No. 740681
NucleoVac 96 Vacuum Manifold vacuum manifold for use of NucleoSpin 8/96 kits under vacuum - manifold base, manifold lid, waste containers, spacer set