Learn More
Macherey-Nagel NucleoVac Vacuum Regulator
Catalog No. NC1031462
Encompass_Preferred
NucleoVac Vacuum Regulator for controlling of vacuum with Vacuum Manifolds, e.g. NucleoVac 24 Vacuum Manifold or NucleoVac 96 Vacuum Manifold
Catalog No. NC1031462 Supplier Macherey-Nagel Supplier No. 740641
NucleoVac Vacuum Regulator for controlling of vacuum with Vacuum Manifolds, e.g. NucleoVac 24 Vacuum Manifold or NucleoVac 96 Vacuum Manifold