Learn More
Macherey-Nagel HPLC column EC 75/4.6 NUCLEODUR C18 Gravity, 3 µm
Supplier:  Macherey-Nagel 760086.46
Encompass_Preferred
EC HPLC column - EC 75/4.6 NUCLEODUR C18 Gravity, 3 µm - length: 75 mm, ID: 4.6 mm - pack of 1
Catalog No. NC1682624