Learn More
Biotium NucSpot 750/780 Nuclear Stain, 100 uL, 1000X in DMSO, Excitation 757/780 nm, Cell membrane-impermeant, Fixed cell staining
Supplier:  Biotium 41038
Encompass_Preferred
NucSpot 750/780 Nuclear Stain, 100 uL, 1000X in DMSO, Excitation 757/780 nm, Cell membrane-impermeant, Fixed cell staining
Catalog No. NC2481349