Learn More
Biotium NucSpot Far-Red, 1000X in DMSO
Catalog No. NC1665614
Encompass_Preferred
NucSpot Far-Red is a dead cell nuclear stain for the PE-Cy5 channel in flow cytometry. NucSpot Far-Red is brighter than the spectrally similar dye 7-AAD in both the PE-Cy5 and APC channels, with less bleed-through into the PE-Texas Red channel, making it more versatile for multiplexing.
Catalog No. NC1665614 Supplier Biotium Supplier No. 40085T
NucSpot Far-Red is a dead cell nuclear stain for the PE-Cy5 channel in flow cytometry. NucSpot Far-Red is brighter than the spectrally similar dye 7-AAD in both the PE-Cy5 and APC channels, with less bleed-through into the PE-Texas Red channel, making it more versatile for multiplexing.