Learn More
Biotium NucSpot Live 488, 50 uL
Catalog No. NC1646780
Encompass_Preferred
NucSpot Live Cell Nuclear Stains are cell-membrane permeable DNA dyes that specifically stains nuclei in live or fixed cells. They have excellent specificity for DNA without the need for a wash step, and they have low toxicity for live cell imaging.
Catalog No. NC1646780 Supplier Biotium Supplier No. 40081
NucSpot Live Cell Nuclear Stains are cell-membrane permeable DNA dyes that specifically stains nuclei in live or fixed cells. They have excellent specificity for DNA without the need for a wash step, and they have low toxicity for live cell imaging.