Learn More
Biotium NucSpot Live 650, 1000X in DMSO
Supplier:  Biotium 40082
Encompass_Preferred
NucSpot Live 650 Live Cell Nuclear Stain is a far-red fluorescent (Ex/Em 650/675 nm) DNA dye that stains nuclei in live or fixed cells. It has low toxicity for live cell imaging and is compatible with super-resolution imaging by STED and SIM. The Supplied as a 1000X stock solution in DMSO.
Catalog No. NC1646782