Learn More
Biotium NucView 530 Caspase-3 Substrate, 1 mM in DMSO
Catalog No. NC1156273
Encompass_Preferred
NucView 530 Caspase-3 Substrate, 1 mM in DMSO, provides a convenient tool for detecting caspase activity in intact cells in real time by either fluorescence microscopy or flow cytometry; the substrate stains apoptotic cell nuclei with orange fluorescence (Ex/Em 528/563 nm).
Catalog No. NC1156273 Supplier Biotium Supplier No. 10406
NucView 530 Caspase-3 Substrate, 1 mM in DMSO, provides a convenient tool for detecting caspase activity in intact cells in real time by either fluorescence microscopy or flow cytometry; the substrate stains apoptotic cell nuclei with orange fluorescence (Ex/Em 528/563 nm).