Learn More
Sino Biological NUDC Antibody, Rabbit PAb, Antigen Affinity Purified 100uL
Supplier:  Sino Biological 15068T62100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human NUDC (rh NUDC; Catalog#15068-H08H; NP_006591.1; Met1-Asn331). NUDC specific IgG was purified by Human NUDC affinity chromatography.
Catalog No. 50-224-8758